Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-62
Entrez Gene ID
175591
Gene Name
UDP-glucuronosyltransferase
Gene Aliases
CELE_M88.1
Chromosome Location
Chr.III: 4537277 - 4540807 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065517.5 NP_497918.1 3 - 4 318 118

View Full Product Details