Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-6
Entrez Gene ID
192070
Gene Name
TWiK family of potassium channels
Gene Aliases
CELE_F17C8.5
Chromosome Location
Chr.III: 4742920 - 4744719 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065572.2 NP_497973.1 3 - 4 592 71

View Full Product Details