Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
col-89
Entrez Gene ID
175627
Gene Name
Col_cuticle_N domain-containing protein
Gene Aliases
CELE_F17C8.2
Chromosome Location
Chr.III: 4748127 - 4749164 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065574.4 NP_497975.1 1 - 2 159 114

View Full Product Details