Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-6
Entrez Gene ID
183242
Gene Name
C-type LECtin
Gene Aliases
CELE_C35D10.15
Chromosome Location
Chr.III: 4885660 - 4887444 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065622.3 NP_498023.1 3 - 4 465 91

View Full Product Details