Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
set-2
Entrez Gene ID
175662
Gene Name
protein-coding
Gene Aliases
CELE_C26E6.9
Chromosome Location
Chr.III: 4922409 - 4930204 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065638.6 NP_498039.1 7 - 8 1441 112
NM_065639.6 NP_498040.1 7 - 8 1514 112

View Full Product Details