Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pqe-1
Entrez Gene ID
175731
Gene Name
protein-coding
Gene Aliases
CELE_F52C9.8
Chromosome Location
Chr.III: 5311240 - 5326509 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047389.4 NP_001040854.1 4 - 5 293 68
NM_065735.6 NP_498136.2 4 - 5 294 68

View Full Product Details