Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dnj-24
Entrez Gene ID
175745
Gene Name
J domain-containing protein
Gene Aliases
CELE_W03A5.7
Chromosome Location
Chr.III: 5415583 - 5424083 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065754.6 NP_498155.3 4 - 5 767 71

View Full Product Details