Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ucr-1
Entrez Gene ID
175772
Gene Name
protein-coding
Gene Aliases
CELE_F56D2.1
Chromosome Location
Chr.III: 5592135 - 5594401 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001356762.3 NP_001343795.1 4 - 4 589 101
NM_001379716.1 NP_001368667.1 1 - 2 64 101
NM_001379717.1 NP_001368666.1 1 - 2 64 101
NM_065801.8 NP_498202.1 4 - 5 612 101

View Full Product Details