Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ucr-1
Entrez Gene ID
175772
Gene Name
protein-coding
Gene Aliases
CELE_F56D2.1
Chromosome Location
Chr.III: 5592135 - 5594401 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001356762.3 NP_001343795.1 9 - 9 1358 75
NM_001379716.1 NP_001368667.1 6 - 7 833 75
NM_001379717.1 NP_001368666.1 6 - 7 827 75
NM_065801.8 NP_498202.1 9 - 10 1375 75

View Full Product Details