Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ceh-10
Entrez Gene ID
175811
Gene Name
Homeobox protein ceh-10
Gene Aliases
CELE_W03A3.1
Chromosome Location
Chr.III: 5799047 - 5801115 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065850.3 NP_498251.1 2 - 3 267 75

View Full Product Details