Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rabn-5
Entrez Gene ID
175823
Gene Name
FYVE-type domain-containing protein
Gene Aliases
CELE_F01F1.4
Chromosome Location
Chr.III: 5864553 - 5866543 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065865.6 NP_498266.1 1 - 2 59 68

View Full Product Details