Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
abcf-3
Entrez Gene ID
175873
Gene Name
ABC transporter, class F
Gene Aliases
CELE_F42A10.1
Chromosome Location
Chr.III: 6173036 - 6176191 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065938.7 NP_498339.1 3 - 4 1081 97

View Full Product Details