Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-7
Entrez Gene ID
191833
Gene Name
Serpentine receptor class gamma-7
Gene Aliases
CELE_C18F10.8
Chromosome Location
Chr.III: 6266484 - 6268161 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065970.2 NP_498371.1 4 - 5 848 108

View Full Product Details