Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sdz-15
Entrez Gene ID
184905
Gene Name
Skn-1 dependent zygotic transcript 15 protein
Gene Aliases
CELE_F23F12.4
Chromosome Location
Chr.III: 6498476 - 6500110 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066031.3 NP_498432.1 2 - 3 1099 133

View Full Product Details