The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001027395.5 | NP_001022566.1 | 12 - 13 | 1993 | 75 |
NM_001027396.5 | NP_001022567.1 | 12 - 13 | 1795 | 75 | |
NM_001379772.1 | NP_001367825.1 | 14 - 15 | 2506 | 75 | |
NM_066066.7 | NP_498467.3 | 14 - 15 | 2469 | 75 | |
GenBank mRNA | AF399852.1 | - | 11 - 12 | 1773 | 75 |