Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cra-1
Entrez Gene ID
175956
Gene Name
N-terminal acetyltransferase B complex subunit NAA25 homolog
Gene Aliases
CELE_R13F6.10
Chromosome Location
Chr.III: 6865206 - 6869896 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066093.6 NP_498494.2 3 - 4 325 107

View Full Product Details