Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
egg-2
Entrez Gene ID
187510
Gene Name
LDL receptor repeat-containing protein egg-2
Gene Aliases
CELE_R01H2.3
Chromosome Location
Chr.III: 7058516 - 7060583 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066136.6 NP_498537.1 2 - 3 132 117

View Full Product Details