Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cup-5
Entrez Gene ID
176074
Gene Name
protein-coding
Gene Aliases
CELE_R13A5.1
Chromosome Location
Chr.III: 7584181 - 7591581 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027550.4 NP_001022721.2 3 - 4 700 114
NM_001027551.5 NP_001022722.2 2 - 3 640 114
NM_001268016.4 NP_001254945.1 2 - 3 640 114
NM_001382919.1 2 - 3 649 114
NM_066263.6 NP_498664.3 2 - 3 635 114
GenBank mRNA AF482952.1 - 2 - 3 634 114

View Full Product Details