Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ncl-1
Entrez Gene ID
176084
Gene Name
protein-coding
Gene Aliases
CELE_ZK112.2
Chromosome Location
Chr.III: 7719725 - 7738105 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001312923.2 NP_001299852.1 8 - 9 1468 116
NM_001312924.2 NP_001299853.1 8 - 9 1411 116
NM_001312995.3 NP_001299924.1 7 - 8 1375 116
NM_001312997.3 NP_001299926.1 6 - 7 1213 116
NM_001312998.3 NP_001299927.1 6 - 7 1204 116
NM_001312999.3 NP_001299928.1 5 - 6 1027 116
NM_001379801.1 NP_001368538.1 7 - 8 1366 116
NM_001379802.1 NP_001367412.1 7 - 8 1383 116
GenBank mRNA AF047027.1 - 5 - 6 1152 116

View Full Product Details