Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
snr-5
Entrez Gene ID
176103
Gene Name
Probable small nuclear ribonucleoprotein F
Gene Aliases
CELE_ZK652.1
Chromosome Location
Chr.III: 7862070 - 7862582 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066307.6 NP_498708.1 1 - 2 97 90

View Full Product Details