Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pcp-5
Entrez Gene ID
176111
Gene Name
Prolyl carboxy peptidase like protein 5
Gene Aliases
CELE_ZK688.6
Chromosome Location
Chr.III: 7904943 - 7907615 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001379810.2 NP_001367399.1 5 - 6 701 104

View Full Product Details