Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gly-3
Entrez Gene ID
176112
Gene Name
Polypeptide N-acetylgalactosaminyltransferase 3
Gene Aliases
CELE_ZK688.8
Chromosome Location
Chr.III: 7914306 - 7917767 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066321.7 NP_498722.1 3 - 4 693 70
GenBank mRNA AF031833.1 - 3 - 4 694 70

View Full Product Details