Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-86
Entrez Gene ID
176157
Gene Name
protein-coding
Gene Aliases
CELE_C30A5.7
Chromosome Location
Chr.III: 8212740 - 8216336 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026020.7 NP_001021191.2 5 - 6 968 71
NM_001322687.4 NP_001309473.1 4 - 5 760 71

View Full Product Details