Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
asna-1
Entrez Gene ID
176254
Gene Name
ATPase asna-1
Gene Aliases
CELE_ZK637.5
Chromosome Location
Chr.III: 8895082 - 8896654 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066564.4 NP_498965.1 3 - 4 900 88

View Full Product Details