Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cls-2
Entrez Gene ID
176280
Gene Name
protein-coding
Gene Aliases
CELE_R107.6
Chromosome Location
Chr.III: 9055048 - 9058899 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276794.3 NP_001263723.1 1 - 2 98 104
NM_066604.5 NP_499005.2 1 - 2 94 104

View Full Product Details