Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lin-12
Entrez Gene ID
176282
Gene Name
Protein lin-12
Gene Aliases
CELE_R107.8
Chromosome Location
Chr.III: 9060096 - 9071472 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066606.6 NP_499007.1 7 - 8 1979 91

View Full Product Details