Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ttr-5
Entrez Gene ID
176312
Gene Name
Transthyretin-like protein 5
Gene Aliases
CELE_C40H1.5
Chromosome Location
Chr.III: 9330316 - 9330928 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066653.4 NP_499054.1 2 - 3 285 115

View Full Product Details