Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mut-7
Entrez Gene ID
176347
Gene Name
Exonuclease mut-7
Gene Aliases
CELE_ZK1098.8
Chromosome Location
Chr.III: 9539029 - 9542605 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066704.9 NP_499105.1 3 - 4 1156 115

View Full Product Details