Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rom-2
Entrez Gene ID
183564
Gene Name
Inactive rhomboid-related protein 2
Gene Aliases
CELE_C48B4.2
Chromosome Location
Chr.III: 9593120 - 9595667 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001379864.1 NP_001366679.1 4 - 5 740 104

View Full Product Details