Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-60
Entrez Gene ID
176366
Gene Name
Putative UDP-glucuronosyltransferase ugt-60
Gene Aliases
CELE_C07A9.6
Chromosome Location
Chr.III: 9676320 - 9679051 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025987.5 NP_001021158.1 1 - 2 173 111

View Full Product Details