Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ekl-6
Entrez Gene ID
176419
Gene Name
Uncharacterized protein
Gene Aliases
CELE_T16G12.5
Chromosome Location
Chr.III: 10064337 - 10069201 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066833.5 NP_499234.2 7 - 8 2121 65

View Full Product Details