Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dpy-28
Entrez Gene ID
176509
Gene Name
Condensin complex subunit 1
Gene Aliases
CELE_Y39A1B.3
Chromosome Location
Chr.III: 10763131 - 10777364 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066978.5 NP_499379.2 5 - 6 1522 96

View Full Product Details