Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pha-1
Entrez Gene ID
176543
Gene Name
protein-coding
Gene Aliases
CELE_Y48A6C.5
Chromosome Location
Chr.III: 11131775 - 11136692 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027719.5 NP_001022890.1 3 - 4 568 140
NM_001027720.2 NP_001022891.1 4 - 5 478 140
GenBank mRNA X73845.1 - - 599 140

View Full Product Details