Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mdt-21
Entrez Gene ID
3565572
Gene Name
Mediator of RNA polymerase II transcription subunit 21
Gene Aliases
CELE_C24H11.9
Chromosome Location
Chr.III: 11777621 - 11780128 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026013.3 NP_001021184.1 1 - 2 213 63

View Full Product Details