Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nob-1
Entrez Gene ID
176641
Gene Name
protein-coding
Gene Aliases
CELE_Y75B8A.2
Chromosome Location
Chr.III: 12077564 - 12084316 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001379944.1 NP_001367008.1 1 - 2 180 77
NM_001383007.1 3 - 4 687 77

View Full Product Details