Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mrg-1
Entrez Gene ID
176702
Gene Name
protein-coding
Gene Aliases
CELE_Y37D8A.9
Chromosome Location
Chr.III: 12871029 - 12874656 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129255.5 NP_001122727.1 1 - 2 498 74
NM_067274.6 NP_499675.1 1 - 2 500 74

View Full Product Details