Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
unc-25
Entrez Gene ID
176713
Gene Name
protein-coding
Gene Aliases
CELE_Y37D8A.23
Chromosome Location
Chr.III: 12942333 - 12949884 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027660.3 NP_001022831.1 1 - 2 384 60
NM_067288.5 NP_499689.1 1 - 2 100 60
GenBank mRNA AF109378.1 - 1 - 2 62 60

View Full Product Details