Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
set-16
Entrez Gene ID
176802
Gene Name
Histone-lysine N-methyltransferase
Gene Aliases
CELE_T12D8.1
Chromosome Location
Chr.III: 13643864 - 13656637 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067418.6 NP_499819.3 5 - 6 1391 71

View Full Product Details