Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyk-4
Entrez Gene ID
176815
Gene Name
CYtoKinesis defect
Gene Aliases
CELE_K08E3.6
Chromosome Location
Chr.III: 13768541 - 13771741 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067444.8 NP_499845.1 3 - 4 263 65

View Full Product Details