The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001372683.1 | NP_001359808.1 | 8 - 9 | 999 | 82 |
NM_001372684.2 | NP_001359809.1 | 8 - 9 | 915 | 82 |