Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tcl-2
Entrez Gene ID
176832
Gene Name
protein-coding
Gene Aliases
CELE_Y38C1AA.4
Chromosome Location
Chr.IV: 179083 - 183254 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307376.3 NP_001294305.1 1 - 2 453 62
NM_067474.7 NP_499875.2 2 - 3 516 62
GenBank mRNA AY145133.1 - 2 - 3 613 62

View Full Product Details