Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cogc-8
Entrez Gene ID
176841
Gene Name
Conserved oligomeric Golgi complex subunit 8
Gene Aliases
CELE_R02D3.2
Chromosome Location
Chr.IV: 240451 - 244130 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067482.4 NP_499883.1 8 - 9 1595 81

View Full Product Details