Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kcc-1
Entrez Gene ID
177495
Gene Name
protein-coding
Gene Aliases
CELE_R13A1.2
Chromosome Location
Chr.IV: 7206269 - 7218002 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306917.3 NP_001293846.1 13 - 14 3365 109
NM_001306918.3 NP_001293847.1 12 - 13 3191 109
NM_001306919.3 NP_001293848.1 11 - 12 3095 109
NM_001307143.2 NP_001294072.1 10 - 11 3230 109
NM_001380315.1 NP_001368174.1 10 - 11 3002 109

View Full Product Details