Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glb-23
Entrez Gene ID
187853
Gene Name
GLOBIN domain-containing protein
Gene Aliases
CELE_R13A1.8
Chromosome Location
Chr.IV: 7218049 - 7219498 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068741.2 NP_501142.1 4 - 5 811 125

View Full Product Details