Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
plc-4
Entrez Gene ID
177525
Gene Name
Phosphoinositide phospholipase C
Gene Aliases
CELE_R05G6.8
Chromosome Location
Chr.IV: 7509465 - 7512667 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068812.4 NP_501213.1 3 - 4 793 149

View Full Product Details