Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hcf-1
Entrez Gene ID
177560
Gene Name
human HCF1 related
Gene Aliases
CELE_C46A5.9
Chromosome Location
Chr.IV: 7770770 - 7774275 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068878.5 NP_501279.1 1 - 2 49 89
GenBank mRNA AF072907.1 - 1 - 2 44 89

View Full Product Details