Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mvb-12
Entrez Gene ID
182286
Gene Name
MVB (yeast MultiVesicular Body sorting factor) related
Gene Aliases
CELE_C06A6.3
Chromosome Location
Chr.IV: 7830562 - 7832435 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068901.7 NP_501302.1 3 - 4 329 86

View Full Product Details