Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-33E1
Entrez Gene ID
183602
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_C49C8.4
Chromosome Location
Chr.IV: 8645521 - 8648602 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069079.6 NP_501480.2 7 - 8 904 92

View Full Product Details