Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nhr-11
Entrez Gene ID
177724
Gene Name
protein-coding
Gene Aliases
CELE_ZC410.1
Chromosome Location
Chr.IV: 9070003 - 9073622 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047598.3 NP_001041063.1 4 - 5 454 152
NM_001268430.4 NP_001255359.1 4 - 5 452 152
GenBank mRNA AY305838.1 - 4 - 5 476 152

View Full Product Details