Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- mppb-1
- Entrez Gene ID
-
177725
- Gene Name
- Mitochondrial-processing peptidase subunit beta
- Gene Aliases
-
CELE_ZC410.2
- Chromosome Location
-
Chr.IV: 9074575 - 9076574 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_069175.8 |
NP_501576.2 |
3 - 4 |
210 |
72 |
- Gene Symbol
- fem-3
- Entrez Gene ID
-
177734
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_C01F6.4
- Chromosome Location
-
Chr.IV: 9074671 - 9107610 on Build WBcel235
View Full Product Details